Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clvs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175448.3 | NP_780657.1 | 2 | 703 | ||
GenBank mRNA | AK039308.1 | - | 2 | 704 | ||
AK049896.1 | - | 2 | 264 | |||
AK049988.1 | - | 2 | 264 | |||
AK053797.1 | - | 2 | 726 | |||
AK139409.1 | - | 2 | 778 | |||
AK139868.1 | - | 2 | 756 | |||
AK163214.1 | - | 2 | 724 | |||
AK169009.1 | - | 2 | 873 | |||
BC058539.1 | - | 2 | 710 | 5690124 |