Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam78b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160261.1 | NP_001153733.1 | 2 | 452 | ||
NM_001160262.1 | NP_001153734.1 | 2 | 452 | |||
NM_175461.4 | NP_780670.2 | 2 | 452 | |||
GenBank mRNA | AK047703.1 | - | 2 | 290 | ||
AK144212.1 | - | 2 | 406 | |||
BC066094.1 | - | 2 | 280 | |||
BC132161.1 | - | 3 | 375 | 40130438 | ||
BC132525.1 | - | 2 | 437 | 40130802 | ||
CB575012.1 | - | 602 | ||||
CF530988.1 | - | 2 | 394 |