Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037727.2 | NP_001032816.1 | 8 | 1356 | ||
NM_001286610.1 | NP_001273539.1 | 7 | 1156 | |||
NM_175476.4 | NP_780685.2 | 8 | 1027 | |||
GenBank mRNA | AK037710.1 | - | 8 | 1089 | ||
AK048450.1 | - | 8 | 987 | |||
AK137444.1 | - | 8 | 1015 | |||
AK137526.1 | - | 8 | 964 | |||
AK159598.1 | - | 7 | 1157 | |||
AK159941.1 | - | 7 | 1157 | |||
AK169897.1 | - | 8 | 1333 | |||
AK172878.1 | - | 6 | 686 | |||
BC108400.1 | - | 5 | 637 | |||
BC138751.1 | - | 7 | 935 | 8861773 | ||
BC138752.1 | - | 7 | 935 | 8861774 |