Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp574 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168506.1 | NP_001161978.1 | 2 | 403 | ||
NM_175477.4 | NP_780686.1 | 2 | 458 | |||
GenBank mRNA | AK042074.1 | - | 2 | 421 | ||
AK053744.1 | - | 2 | 344 | |||
AK143997.1 | - | 2 | 404 | |||
AK170243.1 | - | 2 | 309 | |||
BC021833.2 | - | 2 | 403 | |||
BC054064.1 | - | 2 | 359 | |||
BC059044.1 | - | 2 | 329 | 6816316 | ||
BC094354.1 | - | 2 | 382 |