Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbpl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003717.1 | NP_001003717.1 | 4 | 659 | ||
NM_175489.3 | NP_780698.2 | 5 | 696 | |||
GenBank mRNA | AK037158.1 | - | 5 | 696 | ||
AK050244.1 | - | 4 | 660 | |||
BC141383.1 | - | 5 | 395 | 9055996 |