Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4gat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175383.2 | NP_780592.1 | 2 | 1938 | ||
GenBank mRNA | AK050347.1 | - | 2 | 1912 | ||
AK083857.1 | - | 1 | 2135 | |||
AK158899.1 | - | 2 | 1939 | |||
AV344651.1 | - | 1 | 165 | |||
BC069927.1 | - | 2 | 1880 | 4010980 |
Gene Symbol: Brms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC038380.1 | - | 1873 |