Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sephs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175400.6 | NP_780609.4 | 9 | 2665 | ||
GenBank mRNA | AK044971.1 | - | 9 | 2602 | ||
AK047631.1 | - | 4 | 1696 | |||
AK053638.1 | - | 9 | 2700 | |||
AK075655.1 | - | 2 | 1367 | |||
AK151611.1 | - | 1 | 472 | |||
BC065165.1 | - | 9 | 2626 | 6417054 | ||
BC066037.1 | - | 9 | 2664 | 6405661 |