Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adtrp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145875.1 | NP_001139347.1 | 7 | 2051 | ||
NM_175417.4 | NP_780626.1 | 7 | 1972 | |||
GenBank mRNA | AK029025.1 | - | 7 | 1976 |