Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175483.5 | NP_780692.2 | 1 | 2742 | ||
GenBank mRNA | AK053761.1 | - | 1 | 1277 | ||
AK142274.1 | - | 1 | 2743 | |||
BC096472.1 | - | 1 | 1492 | 3501525 |