Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177673.1 | NP_001171144.1 | 3 | 2813 | ||
NM_001177674.1 | NP_001171145.1 | 3 | 2995 | |||
NM_175493.4 | NP_780702.1 | 3 | 3116 | |||
GenBank mRNA | AI604270.1 | - | 1 | 227 | ||
AK036659.1 | - | 2 | 2825 | |||
AK087410.1 | - | 3 | 2808 | |||
AK154649.1 | - | 2 | 2788 | |||
AK155886.1 | - | 2 | 2806 |
Set Membership: |
G-protein coupled receptor |