Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175500.4 | NP_780709.1 | 1, 2 | 582 | ||
GenBank mRNA | AK038547.1 | - | 1, 2 | 547 | ||
AK084223.1 | - | 1, 2 | 526 | |||
AK134807.1 | - | 1, 2 | 583 | |||
BC115712.1 | - | 1, 2 | 374 | 40059645 |