Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam78a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175511.4 | NP_780720.1 | 1 | 916 | ||
GenBank mRNA | AK077626.1 | - | 1 | 920 | ||
AK079514.1 | - | 1 | 917 | |||
AK165375.1 | - | 1284 | ||||
BC137951.1 | - | 1 | 410 | 8860970 | ||
BC137952.1 | - | 1 | 410 | 8860971 |