Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mum1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164630.1 | NP_001158102.1 | 7 | 1413 | ||
NM_001164631.1 | NP_001158103.1 | 4 | 995 | |||
NM_001164632.1 | NP_001158104.1 | 5 | 1156 | |||
NM_001164633.1 | NP_001158105.1 | 5 | 1095 | |||
NM_175541.3 | NP_780750.2 | 6 | 1336 | |||
GenBank mRNA | AK035086.1 | - | 1 | 164 | ||
AK052099.1 | - | 1 | 2019 | |||
BC068178.1 | - | 4 | 1014 | |||
BC096534.1 | - | 1 | 338 | |||
BC120584.1 | - | 1 | 259 | 40129507 | ||
BC137665.1 | - | 1 | 259 | 8860684 |