Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLNS1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293.2 | NP_001284.1 | 3 | 397 | ||
NM_001311199.1 | NP_001298128.1 | 3 | 397 | |||
NM_001311201.1 | NP_001298130.1 | 3 | 397 | |||
GenBank mRNA | AF005422.1 | - | 3 | 335 | ||
AF026003.1 | - | 3 | 305 | |||
AK315259.1 | - | 3 | 391 | |||
BC119634.1 | - | 3 | 354 | 40115653 | ||
BC119635.1 | - | 3 | 354 | 40115655 | ||
BQ643786.1 | - | 3 | 367 | |||
BT019907.1 | - | 3 | 305 | |||
BU538016.1 | - | 3 | 363 | |||
DA004269.1 | - | 3 | 398 | |||
U17899.1 | - | 3 | 379 | |||
U53454.1 | - | 3 | 389 | |||
X91788.1 | - | 3 | 393 |