Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp384 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252083.1 | NP_001239012.1 | 4 | 434 | ||
NM_175557.5 | NP_780766.3 | 4 | 405 | |||
GenBank mRNA | AK086185.1 | - | 4 | 330 | ||
AK156026.1 | - | 4 | 436 | |||
AK172476.1 | - | 4 | 397 | |||
BB662626.1 | - | 4 | 330 | |||
BC064013.1 | - | 4 | 269 | 6515698 |