Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fchsd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175684.4 | NP_783615.2 | 12 | 1126 | ||
GenBank mRNA | AK079461.1 | - | 6 | 598 | ||
AK154732.1 | - | 12 | 1161 | |||
BC052886.1 | - | 12 | 1157 | |||
BC057367.1 | - | 12 | 1126 | 6827858 |