Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35g1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175507.3 | NP_780716.1 | 3 | 789 | ||
GenBank mRNA | AK041618.1 | - | 3 | 789 | ||
AK052365.1 | - | 3 | 693 | |||
AK171987.1 | - | 3 | 721 | |||
AK209992.1 | - | 1 | 137 | |||
BC137663.1 | - | 3 | 800 | 8860682 | ||
BC137664.1 | - | 3 | 800 | 8860683 |