Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Intu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175515.5 | NP_780724.3 | 396 | |||
GenBank mRNA | AK035092.1 | - | 377 | |||
AK040233.1 | - | 832 | ||||
BC125523.1 | - | 343 | 40130038 | |||
BC132387.1 | - | 343 | 40130664 | |||
BY720518.1 | - | 396 |