Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175537.3 | NP_780746.2 | 3 | 823 | ||
GenBank mRNA | AK044467.1 | - | 3 | 825 | ||
AK054096.1 | - | 3 | 505 | |||
AK054529.1 | - | 3 | 526 | |||
DQ192019.1 | - | 1 | 339 |