Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdfy2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175546.4 | NP_780755.2 | 1, 2 | 376 | ||
GenBank mRNA | AK045072.1 | - | 1, 2 | 181 | ||
AK086275.1 | - | 1, 2 | 389 | |||
AK131958.1 | - | 1, 2 | 375 | |||
AK151817.1 | - | 1, 2 | 332 | |||
AK169499.1 | - | 1, 2 | 353 | |||
BC116632.1 | - | 1, 2 | 322 | 40056912 |