Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175552.4 | NP_780761.1 | 2, 3 | 263 | ||
GenBank mRNA | AK050738.1 | - | 2, 3 | 231 | ||
AK050797.1 | - | 2, 3 | 234 | |||
BC063100.1 | - | 2, 3 | 249 | 6332749 | ||
BY125127.1 | - | 2, 3 | 264 |