Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4930579C12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK029815.1 | - | 5 | 357 |
Gene Symbol: Gm10634 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK077175.1 | - | 1068 |