Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npsr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175678.3 | NP_783609.1 | 6 | 661 | ||
GenBank mRNA | AK033957.1 | - | 6 | 662 | ||
AK041451.1 | - | 5 | 560 | |||
AY255574.1 | - | 1 | 46 | |||
AY299387.1 | - | 5 | 597 |
Set Membership: |
G-protein coupled receptor |