Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175500.4 | NP_780709.1 | 8 | 2231 | ||
GenBank mRNA | AK038547.1 | - | 8 | 2196 | ||
BC115712.1 | - | 8 | 2023 | 40059645 |