Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam78a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175511.4 | NP_780720.1 | 2 | 2098 | ||
GenBank mRNA | AK077626.1 | - | 2 | 2105 | ||
AK079514.1 | - | 2 | 2099 | |||
BC137951.1 | - | 2 | 1592 | 8860970 | ||
BC137952.1 | - | 2 | 1592 | 8860971 |