Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175562.3 | NP_780771.1 | 2 | 1697 | ||
GenBank mRNA | AK041076.1 | - | 1 | 1274 | ||
AK049038.1 | - | 2 | 1697 | |||
AK053487.1 | - | 2 | 1694 | |||
AK083949.1 | - | 1 | 344 | |||
BC049787.1 | - | 2 | 1677 | 6511404 |