Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163818.1 | NP_001157290.1 | 3 | 657 | ||
GenBank mRNA | AJ504718.1 | - | 3 | 663 | ||
AK053183.1 | - | 4 | 676 | |||
AK145485.1 | - | 2 | 101 | |||
BC030903.1 | - | 3 | 664 | |||
BC052059.1 | - | 3 | 660 | 5702171 |