Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ints5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176843.3 | NP_789813.1 | 2 | 177 | ||
GenBank mRNA | AK143868.1 | - | 2 | 139 | ||
AK144770.1 | - | 2 | 152 | |||
AK155499.1 | - | 2 | 139 | |||
AK166149.1 | - | 2 | 150 | |||
AK168795.1 | - | 2 | 150 | |||
AK170584.1 | - | 2 | 139 | |||
AK172260.1 | - | 2 | 139 | |||
BC039214.1 | - | 2 | 159 | 4191047 | ||
BY152900.1 | - | 2 | 179 |