Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXADR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207063.1 | NP_001193992.1 | 2, 3 | 430 | ||
NM_001207064.1 | NP_001193993.1 | 2, 3 | 430 | |||
NM_001207066.1 | NP_001193995.1 | 2, 3 | 430 | |||
NM_001338.4 | NP_001329.1 | 2, 3 | 430 | |||
XM_011529476.2 | XP_011527778.1 | 2, 3 | 320 | |||
XM_011529477.2 | XP_011527779.1 | 2, 3 | 320 | |||
XM_011529478.2 | XP_011527780.1 | 2, 3 | 320 | |||
XM_011529479.1 | XP_011527781.1 | 2, 3 | 320 | |||
GenBank mRNA | AF124598.1 | - | 2, 3 | 206 | ||
AK313526.1 | - | 2, 3 | 293 | |||
AY072911.1 | - | 2, 3 | 206 | |||
AY072912.1 | - | 2, 3 | 206 | |||
BC003684.1 | - | 2, 3 | 307 | 3463613 | ||
BC010536.1 | - | 2, 3 | 281 | 3456544 | ||
BM547121.1 | - | 2, 3 | 354 | |||
BT019876.1 | - | 2, 3 | 206 | |||
CV570985.1 | - | 2, 3 | 430 | |||
GU474812.1 | - | 2, 3 | 206 | |||
U90716.1 | - | 2, 3 | 206 | |||
Y07593.1 | - | 2, 3 | 265 |
Gene Symbol: CXADRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024387.1 | 1 | 287 |
Gene Symbol: LOC100287357 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL834140.1 | - | 3 | 412 |