Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170742.1 | NP_001164213.1 | 6 | 550 | ||
NM_001170743.1 | NP_001164214.1 | 6 | 550 | |||
NM_176923.4 | NP_795897.2 | 6 | 550 | |||
GenBank mRNA | AK030644.1 | - | 6 | 431 | ||
AK136303.1 | - | 2 | 102 | |||
AK170910.1 | - | 6 | 549 | |||
AK173301.1 | - | 6 | 513 | |||
BC066082.1 | - | 6 | 535 | 30355918 | ||
EF011622.1 | - | 6 | 535 |