Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrcam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146031.1 | NP_001139503.1 | 6 | 836 | ||
NM_176930.4 | NP_795904.3 | 6 | 836 | |||
GenBank mRNA | AJ543321.1 | - | 6 | 738 | ||
AK045259.1 | - | 5 | 537 | |||
AK048567.1 | - | 7 | 826 | |||
AK087693.1 | - | 6 | 788 | |||
AK122252.1 | - | 6 | 766 | |||
BC055053.1 | - | 5 | 451 |
Set Membership: |
G-protein coupled receptor |