Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX3X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193416.2 | NP_001180345.1 | 2, 3 | 187 | ||
NM_001356.4 | NP_001347.3 | 2, 3 | 1041 | |||
NR_126093.1 | 2, 3 | 1041 | ||||
XM_011543892.2 | XP_011542194.1 | 2, 3 | 218 | |||
GenBank mRNA | AB208983.1 | - | 2, 3 | 139 | ||
AB451220.1 | - | 2, 3 | 96 | |||
AB451343.1 | - | 2, 3 | 96 | |||
AF000982.1 | - | 2, 3 | 952 | |||
AF000983.1 | - | 2, 3 | 952 | |||
AF061337.1 | - | 2, 3 | 171 | |||
AK291153.1 | - | 2, 3 | 187 | |||
AK296906.1 | - | 2, 3 | 187 | |||
AK297159.1 | - | 2, 3 | 187 | |||
AK304661.1 | - | 2, 3 | 230 | |||
BC011819.2 | - | 2, 3 | 166 | 3617040 | ||
BM833870.1 | - | 2, 3 | 187 | |||
U50553.1 | - | 2, 3 | 405 |