Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rapgef5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175930.5 | NP_787126.3 | 15 | 1790 | ||
GenBank mRNA | AK029995.1 | - | 15 | 1791 | ||
AK030016.1 | - | 15 | 1744 | |||
AK043108.1 | - | 3 | 433 | |||
AK083591.1 | - | 5 | 730 | |||
AK122234.1 | - | 3 | 552 | |||
BC046627.1 | - | 3 | 511 | 6494175 |