Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DECR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001359.1 | NP_001350.1 | 1, 2 | 217 | ||
GenBank mRNA | BC105080.1 | - | 1, 2 | 164 | 8144083 | |
BC105082.1 | - | 1, 2 | 164 | 8144085 | ||
L26050.1 | - | 1, 2 | 86 | |||
U49352.1 | - | 1, 2 | 115 |