Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itga11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176922.5 | NP_795896.4 | 11 | 1304 | ||
GenBank mRNA | AK028821.1 | - | 3 | 214 | ||
AK054303.1 | - | 11 | 1300 | |||
AY124460.1 | - | 11 | 1300 | |||
BC058716.1 | - | 11 | 1298 | 6842988 |