Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DKC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142463.2 | NP_001135935.1 | 4 | 426 | ||
NM_001288747.1 | NP_001275676.1 | 4 | 426 | |||
NM_001363.4 | NP_001354.1 | 4 | 426 | |||
NR_110021.1 | 3 | 903 | ||||
NR_110022.1 | 4 | 426 | ||||
NR_110023.1 | 4 | 426 | ||||
GenBank mRNA | AF067008.1 | - | 4 | 262 | ||
AK225077.1 | - | 4 | 290 | |||
AK307672.1 | - | 4 | 315 | |||
BC009928.2 | - | 4 | 404 | 3604732 | ||
BC010015.2 | - | 4 | 228 | 4303933 | ||
DA429806.1 | - | 4 | 412 | |||
HY050100.1 | - | 4 | 427 | |||
JF279874.1 | - | 4 | 249 | |||
KC954523.1 | - | 3 | 726 | |||
KC954524.1 | - | 4 | 249 | |||
KC954525.1 | - | 4 | 249 | |||
U59151.1 | - | 4 | 282 |