Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162917.1 | NP_001156389.1 | 4 | 761 | ||
GenBank mRNA | AK051202.1 | - | 3 | 503 | ||
AK089365.1 | - | 5 | 726 | |||
AK158187.1 | - | 4 | 601 | |||
AK158279.1 | - | 4 | 660 | |||
BC172087.1 | - | 3 | 419 | 9054373 |