Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176971.2 | NP_795945.1 | 3 | 645 | ||
GenBank mRNA | AB232603.1 | - | 1 | 383 | ||
AK034442.1 | - | 3 | 646 | |||
AK049693.1 | - | 3 | 672 | |||
AK161867.1 | - | 3 | 863 | |||
BC107383.2 | - | 3 | 617 | 40057619 |