Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ush1g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176847.3 | NP_789817.1 | 3 | 3167 | ||
GenBank mRNA | AB087502.1 | - | 3 | 3167 |