Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170742.1 | NP_001164213.1 | 11 | 1028 | ||
NM_001170743.1 | NP_001164214.1 | 11 | 1028 | |||
NM_176923.4 | NP_795897.2 | 11 | 1028 | |||
GenBank mRNA | AK030644.1 | - | 11 | 909 | ||
AK083031.1 | - | 1 | 3775 | |||
AK136303.1 | - | 7 | 580 | |||
AK170910.1 | - | 11 | 1027 | |||
AK173301.1 | - | 11 | 991 | |||
BC066082.1 | - | 11 | 1013 | 30355918 | ||
EF011622.1 | - | 11 | 1013 |