Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nt5dc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176968.4 | NP_795942.2 | 12 | 2502 | ||
GenBank mRNA | AK077876.1 | - | 12 | 2503 | ||
AK145666.1 | - | 12 | 2503 | |||
AK164144.1 | - | 12 | 2443 | |||
AK171761.1 | - | 13 | 2492 | |||
BC045529.1 | - | 1 | 1047 | |||
BC098476.1 | - | 12 | 2462 | 30051251 |