Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176972.4 | NP_795946.2 | 7 | 828 | ||
GenBank mRNA | AK030013.1 | - | 7 | 829 | ||
AK035640.1 | - | 6 | 812 | |||
AK173210.1 | - | 3 | 304 | |||
BC114208.1 | - | 6 | 792 | |||
BC139091.1 | - | 7 | 770 | 8862113 |