Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tigar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177003.5 | NP_795977.1 | 3, 4 | 275 | ||
GenBank mRNA | AK036082.1 | - | 3, 4 | 266 | ||
AK078900.1 | - | 3, 4 | 266 | |||
AK145896.1 | - | 3, 4 | 260 | |||
AK163290.1 | - | 3, 4 | 251 | |||
AK165614.1 | - | 3, 4 | 276 | |||
BC147705.1 | - | 3, 4 | 265 | 9087464 | ||
BC147718.1 | - | 3, 4 | 265 | 9087477 |