Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc17a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177016.3 | NP_795990.2 | 3 | 375 | ||
GenBank mRNA | AK033669.1 | - | 3 | 378 | ||
AK050272.1 | - | 3 | 374 | |||
AK162320.1 | - | 3 | 374 | |||
BC147415.1 | - | 3 | 336 | 9056491 | ||
BC147416.1 | - | 3 | 336 | 9056498 |