Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vwc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177033.3 | NP_796007.1 | 2 | 986 | ||
GenBank mRNA | AB292670.1 | - | 1 | 389 | ||
AK033944.1 | - | 2 | 780 | |||
AK044771.1 | - | 2 | 521 | |||
AK141514.1 | - | 2 | 987 | |||
BC120564.1 | - | 1 | 448 | 40129487 | ||
BC137825.1 | - | 1 | 449 | 8860844 | ||
DQ421811.1 | - | 1 | 452 |