Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177002.3 | NP_795976.1 | 5, 6 | 1162 | ||
GenBank mRNA | AF408394.1 | - | 1064 | |||
AK040781.1 | - | 5, 6 | 1165 | |||
AK050438.1 | - | 5, 6 | 1164 | |||
AK052592.1 | - | 6, 7 | 1260 | |||
AK080180.1 | - | 5, 6 | 1164 | |||
BC016496.1 | - | 5, 6 | 1116 | |||
BC050265.1 | - | 5, 6 | 1134 | 5044850 | ||
BC132146.1 | - | 5, 6 | 1100 | 40130423 | ||
BC132336.1 | - | 5, 6 | 1100 | 40130613 |
Gene Symbol: Slc22a28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013820.3 | NP_001013842.2 | 5, 6 | 1168 | ||
GenBank mRNA | AK052719.1 | - | 5, 6 | 1169 | ||
BC139174.1 | - | 5, 6 | 1100 | 8862196 |