Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apba1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177034.3 | NP_796008.2 | 7 | 1835 | ||
GenBank mRNA | AK032261.1 | - | 6 | 599 | ||
AK147583.1 | - | 7 | 1825 | |||
AK147623.1 | - | 7 | 1787 | |||
AK188637.1 | - | 3 | 189 | |||
BC141181.1 | - | 7 | 1653 | 9055793 |