Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cc2d1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177045.3 | NP_796019.1 | 6 | 653 | ||
GenBank mRNA | AK043839.1 | - | 6 | 654 | ||
AK146601.1 | - | 6 | 672 | |||
AK148162.1 | - | 6 | 640 | |||
AK173271.1 | - | 2 | 81 | |||
BC055068.1 | - | 4 | 352 | |||
BC125517.1 | - | 5 | 543 | 40130032 | ||
BC137743.1 | - | 5 | 543 | 8860762 |