Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jph4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177049.5 | NP_796023.2 | 3 | 2195 | ||
GenBank mRNA | AB096263.1 | - | 1 | 638 | ||
AB099712.1 | - | 3 | 1538 | |||
AK031092.1 | - | 2 | 1242 | |||
AK033956.1 | - | 3 | 1831 | |||
AK034118.1 | - | 3 | 1829 | |||
AK036161.1 | - | 3 | 1841 | |||
AK048607.1 | - | 2 | 1296 | |||
AK148135.1 | - | 2 | 1359 | |||
AY341261.1 | - | 3 | 2195 | |||
AY350744.1 | - | 2 | 1182 | |||
BC052049.1 | - | 2 | 1240 | 5689347 | ||
BC052376.1 | - | 3 | 1292 | 5696231 | ||
BC052722.1 | - | 3 | 1266 | 6810400 |