Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELAVL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001420.3 | NP_001411.2 | 2 | 544 | ||
NM_032281.2 | NP_115657.2 | 2 | 544 | |||
XM_011527778.2 | XP_011526080.1 | 2 | 547 | |||
GenBank mRNA | AY034002.1 | - | 1 | 133 | ||
AY036909.1 | - | 2 | 181 | |||
BC014144.2 | - | 2 | 537 | |||
D26158.1 | - | 2 | 410 | |||
L26405.1 | - | 2 | 310 |